COMH4211BA6D Combi-Blocks, Inc.

1-(Chloromethyl)-2-fluoro-5-methoxy-4-nitrobenzene

  • MDL number MFCD31583121
  • Cas Number : 1257997-14-0
  • Molecular Weight : 219.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6266

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6266
Availability details for
Quantity
  • Availability
  • Add to Cart