COMH4211BA6E Combi-Blocks, Inc.

1-[3-(2-Methoxy-ethoxy)-phenyl]-ethanone

  • MDL number MFCD09943404
  • Cas Number : 73244-53-8
  • Molecular Weight : 194.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6270

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6270
Availability details for
Quantity
  • Availability
  • Add to Cart