COMH4211BA6F Combi-Blocks, Inc.

2,2-Difluoropropyl 4-nitrobenzenesulfonate

  • MDL number MFCD28383645
  • Cas Number : 1268475-14-4
  • Molecular Weight : 281.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6273

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6273
Availability details for
Quantity
  • Availability
  • Add to Cart