COMH4211BA72 Combi-Blocks, Inc.

4-Bromo-3-methyl-n-(2-methylpropyl)benzamide

  • MDL number MFCD26416042
  • Cas Number : 1707600-24-5
  • Molecular Weight : 270.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6277

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6277
Availability details for
Quantity
  • Availability
  • Add to Cart