COMH4211BA73 Combi-Blocks, Inc.

4-Hydroxy-n,n,2-trimethylbenzamide

  • MDL number MFCD16997913
  • Cas Number : 1243472-58-3
  • Molecular Weight : 179.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6278

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6278
Availability details for
Quantity
  • Availability
  • Add to Cart