COMH4211BA79 Combi-Blocks, Inc.

4-(4-Formyl-phenoxy)-piperidine-1-carboxylic acid benzyl ester

  • MDL number MFCD26385701
  • Cas Number : 199103-20-3
  • Molecular Weight : 339.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6292

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6292
Availability details for
Quantity
  • Availability
  • Add to Cart