COMH4211BA7A Combi-Blocks, Inc.

3-(4-Iodo-phenoxy)-2,2-dimethyl-propionic acid

  • MDL number MFCD23731648
  • Cas Number : 1517999-55-1
  • Molecular Weight : 320.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6293

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6293
Availability details for
Quantity
  • Availability
  • Add to Cart