COMH4211BA7B Combi-Blocks, Inc.

tert-Butyl (2-bromo-4-trifluoromethylphenoxy)acetate

  • MDL number MFCD26395159
  • Cas Number : 779331-60-1
  • Molecular Weight : 355.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6295

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6295
Availability details for
Quantity
  • Availability
  • Add to Cart