COMH4211BA7E Combi-Blocks, Inc.

Nbd-x, se

  • MDL number MFCD00467955
  • Cas Number : 145195-58-0
  • Molecular Weight : 391.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6299

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6299
Availability details for
Quantity
  • Availability
  • Add to Cart