COMH4211BA80 Combi-Blocks, Inc.

4-(2-Methyl-benzyloxymethyl)-piperidine hydrochloride

  • MDL number MFCD18837529
  • Cas Number : 1289384-88-8
  • Molecular Weight : 255.8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6302

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6302
Availability details for
Quantity
  • Availability
  • Add to Cart