COMH4211BA82 Combi-Blocks, Inc.

Methyl 2,2-dimethyl-3-oxo-3,4-dihydro-2h-1,4-benzoxazine-6-carboxylate

  • MDL number MFCD26747244
  • Cas Number : 179950-75-5
  • Molecular Weight : 235.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6307

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6307
Availability details for
Quantity
  • Availability
  • Add to Cart