COMH4211BA83 Combi-Blocks, Inc.

4-Amino-n-(cyclopropylmethyl)benzamide

  • MDL number MFCD09734706
  • Cas Number : 934524-22-8
  • Molecular Weight : 190.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6315

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6315
Availability details for
Quantity
  • Availability
  • Add to Cart