COMH4211BA86 Combi-Blocks, Inc.

4-(4-Benzyloxycarbonyl-3-fluoro-phenyl)-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31579528
  • Cas Number : 2270907-69-0
  • Molecular Weight : 414.5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6324

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6324
Availability details for
Quantity
  • Availability
  • Add to Cart