COMH4211BA8A Combi-Blocks, Inc.

2-[2-(2,2,2-Trifluoro-ethoxy)-phenyl]-ethanol

  • MDL number MFCD31628783
  • Cas Number : 1691607-26-7
  • Molecular Weight : 220.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6340

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6340
Availability details for
Quantity
  • Availability
  • Add to Cart