COMH4211BA8B Combi-Blocks, Inc.

(3,3-Dimethyl-piperidin-4-yl)-acetic acid hydrochloride

  • MDL number MFCD31579389
  • Cas Number : 2270905-30-9
  • Molecular Weight : 207.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6341

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6341
Availability details for
Quantity
  • Availability
  • Add to Cart