COMH4211BA8E Combi-Blocks, Inc.

2-(4-Bromo-2,6-difluoro-phenoxy)-2-methyl-propionic acid

  • MDL number MFCD31628748
  • Cas Number : 2168634-16-8
  • Molecular Weight : 295.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6347

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6347
Availability details for
Quantity
  • Availability
  • Add to Cart