COMH4211BA8F Combi-Blocks, Inc.

2-(2-Amino-5-bromo-pyridin-3-yl)-propan-2-ol

  • MDL number MFCD30291787
  • Cas Number : 304467-35-4
  • Molecular Weight : 231.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6349

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6349
Availability details for
Quantity
  • Availability
  • Add to Cart