COMH4211BA90 Combi-Blocks, Inc.

1-(2-Fluoro-4-methyl-phenyl)-ethylamine

  • MDL number MFCD18708687
  • Cas Number : 1270514-56-1
  • Molecular Weight : 153.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6352

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6352
Availability details for
Quantity
  • Availability
  • Add to Cart