COMH4211BA92 Combi-Blocks, Inc.

3-Bromo-5-(propane-2-sulfonylamino)-benzoic acid

  • MDL number MFCD25137851
  • Cas Number : 1515323-52-0
  • Molecular Weight : 322.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6355

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6355
Availability details for
Quantity
  • Availability
  • Add to Cart