COMH4211BAE1 Combi-Blocks, Inc.

5-Bromo-2-cyclopropylmethoxy-3-fluoro-benzaldehyde

  • MDL number MFCD31583169
  • Cas Number : 2243186-78-7
  • Molecular Weight : 273.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6358

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6358
Availability details for
Quantity
  • Availability
  • Add to Cart