COMH4211BAE4 Combi-Blocks, Inc.

[(3,5-Difluorophenyl)methyl](propan-2-yl)amine

  • MDL number MFCD11212514
  • Cas Number : 1094512-00-1
  • Molecular Weight : 185.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6364

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6364
Availability details for
Quantity
  • Availability
  • Add to Cart