COMH4211BAE5 Combi-Blocks, Inc.

6-Bromo-n-ethyl-3h-imidazo[4,5-b]pyridin-2-amine

  • MDL number MFCD26389575
  • Cas Number : 1256957-80-8
  • Molecular Weight : 241.09

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromo-N-ethyl-3H-imidazo[4,5-b]pyridin-2-amine
Inchi Code 1S/C8H9BrN4/c1-2-10-8-12-6-3-5(9)4-11-7(6)13-8/h3-4H,2H2,1H3,(H2,10,11,12,13)
Inchi Key GIMQOOGPHZDCPL-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6367

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6367
Availability details for
Quantity
  • Availability
  • Add to Cart