COMH4211BAE8 Combi-Blocks, Inc.

6-(1H-1,2,3,4-Tetrazol-5-yl)pyridin-3-amine

  • MDL number MFCD27932090
  • Cas Number : 908581-41-9
  • Molecular Weight : 162.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6374

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6374
Availability details for
Quantity
  • Availability
  • Add to Cart