COMH4211BAE9 Combi-Blocks, Inc.

Benzamide, n-[2-[2-(2-aminoethoxy)ethoxy]ethyl]-4-iodo-

  • MDL number MFCD30578230
  • Cas Number : 669077-51-4
  • Molecular Weight : 378.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6376

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6376
Availability details for
Quantity
  • Availability
  • Add to Cart