COMH4211BAEA Combi-Blocks, Inc.

1-(3-Aminophenyl)-3-o-tolylurea

  • MDL number MFCD11123691
  • Cas Number : 1036431-96-5
  • Molecular Weight : 241.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6377

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6377
Availability details for
Quantity
  • Availability
  • Add to Cart