COMH4211BAEC Combi-Blocks, Inc.

4-Fluoro-2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1h-pyrazol-1-yl)methyl)benzonitrile

  • MDL number MFCD28718202
  • Cas Number : 1604036-96-5
  • Molecular Weight : 327.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-fluoro-2-((4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrazol-1-yl)methyl)benzonitrile
Inchi Code 1S/C17H19BFN3O2/c1-16(2)17(3,4)24-18(23-16)14-9-21-22(11-14)10-13-7-15(19)6-5-12(13)8-20/h5-7,9,11H,10H2,1-4H3
Inchi Key AWNHLXYKFVQQJJ-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6380

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6380
Availability details for
Quantity
  • Availability
  • Add to Cart