COMH4211BAEF Combi-Blocks, Inc.

Sodium 2-(2-bromo-phenyl)ethanesulfonate

  • MDL number MFCD31583283
  • Cas Number : 1033629-58-1
  • Molecular Weight : 287.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6384

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6384
Availability details for
Quantity
  • Availability
  • Add to Cart