COMH4211BAF0 Combi-Blocks, Inc.

4-(3,3-Difluoro-pyrrolidin-1-yl)-butyric acid hydrochloride

  • MDL number MFCD31613850
  • Cas Number : 2270905-94-5
  • Molecular Weight : 229.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6387

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6387
Availability details for
Quantity
  • Availability
  • Add to Cart