COMH4211BAF1 Combi-Blocks, Inc.

6-(Ethylsulfonylamino)hexanoic acid

  • MDL number MFCD09728035
  • Cas Number : 844681-28-3
  • Molecular Weight : 223.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6388

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6388
Availability details for
Quantity
  • Availability
  • Add to Cart