COMH4211BAF2 Combi-Blocks, Inc.

2-(4-Aminophenyl)-n-(cyclopropylmethyl)acetamide

  • MDL number MFCD09734363
  • Cas Number : 953883-50-6
  • Molecular Weight : 204.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6392

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6392
Availability details for
Quantity
  • Availability
  • Add to Cart