COMH4211BAF3 Combi-Blocks, Inc.

2'-Fluoro-3-methylbiphenyl-4-carboxylic acid

  • MDL number MFCD14701524
  • Cas Number : 1008773-85-0
  • Molecular Weight : 230.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6394

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6394
Availability details for
Quantity
  • Availability
  • Add to Cart