COMH4211BAF9 Combi-Blocks, Inc.

4-(4-Carbamoyl-3-fluoro-phenyl)-2-methyl-piperazine-1-carboxylic acid tert-butyl ester

  • MDL number MFCD31579535
  • Cas Number : 2270908-27-3
  • Molecular Weight : 337.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6413

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6413
Availability details for
Quantity
  • Availability
  • Add to Cart