COMH4211BAFA Combi-Blocks, Inc.

4-Bromo-n-cyclopropylmethyl-3-fluorobenzamide

  • MDL number MFCD16084879
  • Cas Number : 515135-58-7
  • Molecular Weight : 272.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-N-(cyclopropylmethyl)-3-fluorobenzamide
Inchi Code 1S/C11H11BrFNO/c12-9-4-3-8(5-10(9)13)11(15)14-6-7-1-2-7/h3-5,7H,1-2,6H2,(H,14,15)
Inchi Key IHPRNFONJPGMPS-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6414

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6414
Availability details for
Quantity
  • Availability
  • Add to Cart