COMH4211BAFB Combi-Blocks, Inc.

Hexanoic acid, 6-[(triphenylmethyl)thio]-, 2,5-dioxo-1-pyrrolidinyl ester

  • MDL number MFCD31381700
  • Cas Number : 1398623-71-6
  • Molecular Weight : 487.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6415

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6415
Availability details for
Quantity
  • Availability
  • Add to Cart