COMH4211BAFC Combi-Blocks, Inc.

1-(2,5-Dichlorophenyl)-2,2,2-trifluoroethanol

  • MDL number MFCD12787834
  • Cas Number : 1249336-72-8
  • Molecular Weight : 245.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6417

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6417
Availability details for
Quantity
  • Availability
  • Add to Cart