COMH4211BAFD Combi-Blocks, Inc.

3-Methoxymethoxy-2,2-dimethylpropionic acid chloromethyl ester

  • MDL number MFCD31583273
  • Cas Number : 1350551-93-7
  • Molecular Weight : 210.7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6419

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6419
Availability details for
Quantity
  • Availability
  • Add to Cart