COMH4211BAFE Combi-Blocks, Inc.

1-(2-(tert-Butyldimethylsilyloxy)ethyl)-1h-pyrazol-4-amine

  • MDL number MFCD26393132
  • Cas Number : 1346672-87-4
  • Molecular Weight : 241.41

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6421

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6421
Availability details for
Quantity
  • Availability
  • Add to Cart