COMH4211BB00 Combi-Blocks, Inc.

4-Bromo-3-(2,2,2-trifluoroethoxy)benzamide

  • MDL number MFCD30830137
  • Cas Number : 1443763-87-8
  • Molecular Weight : 298.06

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-3-(2,2,2-trifluoroethoxy)benzamide
Inchi Code 1S/C9H7BrF3NO2/c10-6-2-1-5(8(14)15)3-7(6)16-4-9(11,12)13/h1-3H,4H2,(H2,14,15)
Inchi Key CFRDJNOBCVXNRR-UHFFFAOYSA-N

Description

Purity 95%
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6424

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6424
Availability details for
Quantity
  • Availability
  • Add to Cart