COMH4211BB01 Combi-Blocks, Inc.

4-Amino-3-(2-methoxyethoxy)-n,n-dimethylbenzamide

  • MDL number MFCD18566203
  • Cas Number : 1493977-58-4
  • Molecular Weight : 238.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin USA
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6425

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6425
Availability details for
Quantity
  • Availability
  • Add to Cart