COMH4211BB02 Combi-Blocks, Inc.

4-(3-(Trifluoromethyl)phenyl)-1h-indole

  • MDL number MFCD18422366
  • Cas Number : 442562-70-1
  • Molecular Weight : 261.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6426

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6426
Availability details for
Quantity
  • Availability
  • Add to Cart