COMH4211BB07 Combi-Blocks, Inc.

2-(Azetidin-1-yl)-5-bromopyridine

  • MDL number MFCD23703056
  • Cas Number : 708273-64-7
  • Molecular Weight : 213.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6433

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6433
Availability details for
Quantity
  • Availability
  • Add to Cart