COMH4211BB09 Combi-Blocks, Inc.

1-(2-Cyclopropylmethoxy-4-fluoro-phenyl)-ethanone

  • MDL number MFCD17286510
  • Cas Number : 1343853-62-2
  • Molecular Weight : 208.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6442

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6442
Availability details for
Quantity
  • Availability
  • Add to Cart