COMH4211BB0C Combi-Blocks, Inc.

1-(4-(1H-Pyrrol-1-yl)phenyl)propan-1-one

  • MDL number MFCD00828978
  • Cas Number : 245728-56-7
  • Molecular Weight : 199.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6448

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6448
Availability details for
Quantity
  • Availability
  • Add to Cart