COMH4211BB0D Combi-Blocks, Inc.

2-Chloro-6-(3-chloro-4-fluoro-phenyl)-isonicotinic acid methyl ester

  • MDL number MFCD31613694
  • Cas Number : 1569088-22-7
  • Molecular Weight : 300.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin CN
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6449

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6449
Availability details for
Quantity
  • Availability
  • Add to Cart