COMH4211BB0F Combi-Blocks, Inc.

N-Methoxy-n-methylnaphthalene-1-carboxamide

  • MDL number MFCD10569141
  • Cas Number : 226931-26-6
  • Molecular Weight : 215.2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6453

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6453
Availability details for
Quantity
  • Availability
  • Add to Cart