COMH4211BB11 Combi-Blocks, Inc.

(4-[(4-Methylphenyl)methoxy]phenyl)methanol

  • MDL number MFCD09375894
  • Cas Number : 77350-58-4
  • Molecular Weight : 228.3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6455

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6455
Availability details for
Quantity
  • Availability
  • Add to Cart