COMH4211BB13 Combi-Blocks, Inc.

Aripiprazole 3-deschloro impurity

  • MDL number MFCD31382414
  • Cas Number : 203395-81-7
  • Molecular Weight : 413.9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6464

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6464
Availability details for
Quantity
  • Availability
  • Add to Cart