COMH4211BB14 Combi-Blocks, Inc.

3-Iodo-4-methoxy-1h-pyrazole

  • MDL number MFCD31579305
  • Cas Number : 1533442-08-8
  • Molecular Weight : 224.0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6472

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6472
Availability details for
Quantity
  • Availability
  • Add to Cart