COMH4211BB16 Combi-Blocks, Inc.

2-(2-((tert-Butoxycarbonyl)amino)ethoxy)-ethyl 4-methylbenzenesulfonate

  • MDL number MFCD28506265
  • Cas Number : 192132-77-7
  • Molecular Weight : 359.4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6474

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6474
Availability details for
Quantity
  • Availability
  • Add to Cart