COMH4211BB17 Combi-Blocks, Inc.

5-Bromo-2-(cyclopropylmethoxy)benzamide

  • MDL number MFCD29094337
  • Cas Number : 1369916-26-6
  • Molecular Weight : 270.1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Country of Origin China
Product Link http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6477

Safety Information

MSDS http://www.combi-blocks.com/cgi-bin/find.cgi?QD-6477
Availability details for
Quantity
  • Availability
  • Add to Cart